Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSEKVDWLQ SQNGVCKVDV YSPGDNQAQD WKMDTSTDPV RVLSWLRRDL EKSTAEFQDV RFKPGESFGG ETSNSGDPHK GFSVDYYNTT TKGTPERLHF EMTHKEIPCQ GPRAQLGNES SVDEVSFYAN RLTNLVIAMA RKEINEKIDG SENKCVYQSL YMGNEPTPTK SLSKIASELV NETVSACSRN AAPDKAPGSG DRVSGSSQSP PNLKYKSTLK IKESTKERQG PDDKPPSKKS FFYKEVFESR NGDYAREGGR FFPRERKRFR GQERPDDFTA SVSEGIMTYA NSVVSDMMVS I (1-300 aa encoded by BC047535)
The type II cAMP-dependent protein kinase (PKA) is a multifunctional kinase with a broad range of substrates. Specificity of PKA signaling is mediated by the compartmentalization of the kinase to specific sites within the cell. To maintain this specific localization, the R subunit (RII) of PKA interacts with specific RII-anchoring proteins, designated A-kinase anchoring proteins (AKAP). AKAP 3, also known as AKAP 110, FSP95, PRKA3 and SOB1, binds both PKA and PDE4A and functions as a scaffolding protein in spermatozoa to regulate local cAMP concentrations and modulate sperm functions. Expression of AKAP 3 in normal tissues is restricted to the testis, where bicarbonate stimulates tyrosine phosphorylation of AKAP 3, thereby increasing its recruitment of PKA. AKAP-3 also exhibits high expression in patients with epithelial ovarian cancer (EOC). It demonstrates tumor-restricted expression and appears to be associated with worse overall survival, which make AKAP 3 a potential target for antigen-specific immunotherapy in EOC.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A kinase (PRKA) anchor protein 3; A-kinase anchor protein 110 kDa; A-kinase anchor protein 3; A-kinase anchor protein, 110kDa; AKAP 110; AKAP-3; Cancer/testis antigen 82; CT82; epididymis luminal protein 159; Fibrous Sheath Protein of 95 kDa; fibrousheathin 1; Fibrousheathin I; Fibrousheathin-1; FSP95; PRKA3; protein kinase A binding protein AKAP 110; Protein kinase A-anchoring protein 3; Sperm oocyte-binding protein; sperm oocyte-binding protein 1
Gene Aliases: AKAP 110; AKAP110; AKAP3; CT82; FSP95; HEL159; PRKA3; SOB1
UniProt ID: (Human) O75969, (Mouse) O88987
Entrez Gene ID: (Human) 10566, (Mouse) 11642
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support