Applications | Tested Dilution | Publications |
---|---|---|
Immunohistochemistry (Paraffin) (IHC (P)) |
1:100 | - |
Immunocytochemistry (ICC/IF) |
1:100 | - |
Product Specifications | |
---|---|
Species Reactivity |
Dog, Human |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
UMAB45 |
Immunogen |
Full length human recombinant protein of human ALDH1L1 produced in HEK293T cell. |
Conjugate |
Unconjugated |
Form |
liquid |
Concentration |
0.5-1.0 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS with 1% BSA, 50% glycerol |
Contains |
0.02% sodium azide |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
ALDH1L1 catalyzes the conversion of 10-formyltetrahydrofolate, NADP, and water to tetrahydrofolate, NADPH, and carbon dioxide. The encoded protein belongs to the aldehyde dehydrogenase family and is responsible for formate oxidation in vivo. Deficiencies in this gene can result in an accumulation of formate and subsequent methanol poisoning.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 10-formyltetrahydrofolate dehydrogenase; 10-FTHFDH; 10fTHF; 10FTHFDH; aldehyde dehydrogenase 1 family, member L1; Aldehyde dehydrogenase family 1 member L1; Cytosolic 10-formyltetrahydrofolate dehydrogenase; DKFZp781N0997; MER57B-ALDH1L1
Gene Aliases: 10-fTHF; 10-FTHFDH; ALDH1L1; FDH; FTHFD
UniProt ID: (Human) O75891
Entrez Gene ID: (Human) 10840, (Dog) 100855730
Molecular Function:
RNA processing factor
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support