Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: MSGAPTAGAA LMLCAATAVL LSAQGGPVQS KSPRFASWDE MNVLAHGLLQ LGQGLREHAE RTRSQLSALE RRLSACGSAC QGTEGSTDLP LAPESRVDPE VLHSLQTQLK AQNSRIQQLF HKVAQQQRHL EKQHLRIQHL QSQFGLLDHK HLDHEVAKPA RRKRLPEMAQ PVDPAHNVSR LHRLPRDCQE LFQVGERQSG LFEIQPQGSP PFLVNCKMTS DGGWTVIQRR HDGSVDFNRP WEAYKAGFGD PHGEFWLGLE KVHSITGDRN SRLAVQLRDW DGNAELLQFS VHLGGEDTAY SLQLTAPVAG QLGATTVPPS GLSVPFSTWD QDHDLRRDKN CAKSLSGGWW FGTCSHSNLN GQYFRSIPQQ RQKL
Angiopoietin-like protein 4 (ANGPTL4), is a secreted protein selectively expressed in adipose tissue, liver, and placenta1-3 that plays a variety of roles in vivo, ranging from adipogenesis to angiogenesis to carcinogenesis. The expression of ANGPTL4 is also under nutritional and hormonal control. ANGPTL4 has been observed to induce a strong pro-angiogenic response independent of vascular endothelial growth factor (VEGF), and its expression has been described in hypoxic human tissues as well as a variety of cancers, including liposarcoma, hepatocellular carcinoma, and conventional renal cell carcinoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 425O18-1; angiopoietin-like 4; Angiopoietin-like protein 4; Angiopoietin-related protein 4; ANGPTL2; Fasting-i; Fasting-induced adipose factor; fibrinogen/angiopoietin-related protein; hepatic angiopoietin-related protein; Hepatic fibrinogen/angiopoietin-related protein; HFARP; major histocompatibility complex region NG27; peroxisome proliferator-activated receptor (PPAR) gamma induced angiopoietin-related protein; PPARG angiopoietin related protein; Secreted protein Bk89
Gene Aliases: ANGPTL4; ARP4; Bk89; Farp; FIAF; HARP; HFARP; Ng27; NL2; PGAR; Pgarg; PP1158; PSEC0166; TGQTL; UNQ171; UNQ171/PRO197
UniProt ID: (Human) Q9BY76, (Mouse) Q9Z1P8
Entrez Gene ID: (Human) 51129, (Mouse) 57875
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support