The recommended shelf life for this product is 1 year from date of receipt.
ANKFY1 encodes a cytoplasmic protein that contains a coiled-coil structure and a BTB/POZ domain at its N-terminus, ankyrin repeats in the middle portion, and a FYVE-finger motif at its C-terminus. This protein belongs to a subgroup of double zinc finger proteins which may be involved in vesicle or protein transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat and FYVE domain-containing protein 1; ankyrin repeat hooked to a zinc finger motif; ankyrin repeat hooked to zinc finger motif; Ankyrin repeats hooked to a zinc finger motif; Rabankyrin-5; Rank-5
Gene Aliases: ANKFY1; ANKHZN; BTBD23; KIAA1255; mKIAA1255; ZFYVE14
UniProt ID: (Human) Q9P2R3, (Mouse) Q810B6
Entrez Gene ID: (Human) 51479, (Mouse) 11736
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support