Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MMVLKVEELV TGKKNGNGEA GEFLPEDFRD GEYEAAVTLE KQEDLKTLLA HPVTLGEQQW KSEKQREAEL KKKKLEQRSK LENLEDLEII IQLKKRKKYR KTKVPVVKEP EPEIITEPVD VPTFLKAALE NKLPVVEKFL SDKNNPDVCD EYKRTALHRA CLEGHLAIVE KLMEAGAQIE FRDMLESTAI HWASRGGNLD VLKLLLNKGA KISARDKLLS TALHVAVRTG HYECAEHLIA CEADLNAKDR EGDTPLHDAV RLNRYKMIRL LIMYGADLNI KNCAGKTPMD LVLHWQNGTK AIFDSLRENS YKTSRIATF
The protein encoded by this gene is localized to the nucleus of endothelial cells and is induced by IL-1 and TNF-alpha stimulation. Studies in rat cardiomyocytes suggest that this gene functions as a transcription factor. Interactions between this protein and the sarcomeric proteins myopalladin and titin suggest that it may also be involved in the myofibrillar stretch-sensor system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ankyrin repeat domain 1 (cardiac muscle); Ankyrin repeat domain-containing protein 1; ankyrin-like repeat protein; Cardiac adriamycin-responsive protein; Cardiac ankyrin repeat protein; cardiac responsive adriamycin protein; CARP; Cytokine-inducible gene C-193 protein; Cytokine-inducible nuclear protein; liver ankyrin repeat domain 1; muscle ankyrin repeat protein
Gene Aliases: ALRP; ANKRD1; bA320F15.2; C-193; C193; CARP; Crap; CVARP; HA1A2; MARP; MARP1; MCARP
UniProt ID: (Rabbit) Q9TU71, (Human) Q15327, (Pig) Q865U8, (Mouse) Q9CR42, (Rat) Q8R560
Entrez Gene ID: (Rabbit) 100008719, (Human) 27063, (Pig) 396959, (Mouse) 107765, (Rat) 27064
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support