Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MTGEKIRSL RRDHKPSKEE GDLLEPGDEE AAAALGGTFT RSRIGKGGKA CHKIFSNHHH RLQLKAAPAS SNPPGAPALP LHNSSVTANS QSPALLAGTN PVAVVADGGS CPAHYPVHEC VFKGDVRRLS SLIRTHNIGQ KDNHG (1-144 aa encoded by BC028840)
Acts as a molecular chaperone for G protein-coupled receptors, regulating their biogenesis and exit from the ER.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat domain-containing protein 13C; RP4-677H15.5; unnamed protein product
Gene Aliases: AI505652; ANKRD13C; AU022220; dJ677H15.3
UniProt ID: (Human) Q8N6S4, (Mouse) Q3UX43
Entrez Gene ID: (Human) 81573, (Mouse) 433667
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support