Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Involved in the endosome-to-plasma membrane trafficking and recycling of SNX27-retromer-dependent cargo proteins, such as GLUT1 (PubMed:25278552). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ankrin repeat domain 50; Ankyrin repeat domain-containing protein 50
Gene Aliases: AI662170; ANKRD50; E430012K20Rik; KIAA1223; mKIAA1223; mKIAA3019
UniProt ID: (Human) Q9ULJ7
Entrez Gene ID: (Human) 57182, (Mouse) 99696
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support