Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SOWAHC (Sosondowah Ankyrin Repeat Domain Family Member C) is a Protein Coding gene. An important paralog of this gene is SOWAHA. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ankyrin repeat domain 57; Ankyrin repeat domain-containing protein 57; Ankyrin repeat domain-containing protein SOWAHC; Protein sosondowah homolog C
Gene Aliases: ANKRD57; C2orf26; SOWAHC
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support