Reconstitute antibody with 50 µL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Peptide sequence is: (C)KLPKNIN GTDPIQK
Homology: Mouse, rat - 12/14 amino acid residues identical.
May act as a calcium-activated chloride channel.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Anoctamin-6; SCAN channel; Small-conductance calcium-activated nonselective cation channel; Transmembrane protein 16F
Gene Aliases: 2900059G15Rik; AA407480; ANO6; AW554778; BDPLT7; F730003B03Rik; SCTS; TMEM16F
UniProt ID: (Human) Q4KMQ2, (Mouse) Q6P9J9
Entrez Gene ID: (Human) 196527, (Rat) 315272, (Mouse) 105722
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support