Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MAKGFYISKS LGILGILLGV AAVCTIIALS VVYSQEKNKN ANSSPVASTT PSASATTNPA SATTLDQSKA WNRYRLPNTL KPDSYQVTLR PYLTPNDRGL YVFKGSSTVR FTCKEATDVI IIHSKKLNYT LSQGHRVVLR GVGGSQPPDI DKTELVEPTE YLVVHLKGSL VKDSQYEMDS EFEGELADDL AGFYRSEYME GNVRKVVATT QMQAADARKS FPCFDEPAMK AEFNITLIHP KDLTALSNML PKGPSTPLPE DPNWNVTEFH TTPKMSTYLL AFIVSEFDYV EKQASNGVLI RIWARPSAIA AGHGDYALNV TGPILNFFAG HYDTPYPLPK SDQIGLPDFN AGAMENWGLV TYRENSLLFD PLSSSSSNKE RVVTVIAHEL AHQWFGNLVT IEWWNDLWLN EGFASYVEYL GADYAEPTWN LKDLMVLNDV YRVMAVDALA SSHPLSTPAS EINTPAQISE LFDAISYSKG ASVLRMLSSF LSEDVFKQGL ASYLHTFAYQ NTIYLNLWDH LQEAVNNRSI QLPTTVRDIM NRWTLQMGFP VITVDTSTGT LSQEHFLLDP DSNVTRPSEF NYVWIVPITS IRDGRQQQDY WLIDVRAQND LFSTSGNEWV LLNLNVTGYY RVNYDEENWR KIQTQLQRDH SAIPVINRAQ IINDAFNLAS AHKVPVTLAL NNTLFLIEER QYMPWEAALS SLSYFKLMFD RSEVYGPMKN YLKKQVTPLF IHFRNNTNNW REIPENLMDQ YSEVNAISTA CSNGVPECEE MVSGLFKQWM ENPNNNPIHP NLRSTVYCNA IAQGGEEEWD FAWEQFRNAT LVNEADKLRA ALACSKELWI LNRYLSYTLN PDLIRKQDAT STIISITNNV IGQGLVWDFV QSNWKKLFND YGGGSFSFSN LIQAVTRRFS TEYELQQLEQ FKKDNEETGF GSGTRALEQA LEKTKANIKW VKENKEVVLQ WFTENSK
CD13 (ANPEP) is a 150-170 kDa type II transmembrane zinc-binding ectopeptidase expressed on various cell types. CD13 is a metalloprotease that preferentially catalyzes removal of neutral amino acids from small peptides, thus activating or inactivating bioactive peptides. CD13 also has a role in extracellular matrix degradation, antigen processing and signal transduction, is important in inflammatory responses, regulates intercellular contact, cell motility and vascularization. CD13 is expressed by granulocytes, monocytes, myeloid leukemia cells, and is involved in protection of leukemic cells against apoptosis and its expression associated with poor prognosis of carcinomas. CD13 functions as an aminopeptidase enzyme and is also alpha receptor for coronavirus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alanyl (membrane) aminopeptidase; Alanyl aminopeptidase; Aminopeptidase M; Aminopeptidase N; AP-M; AP-N; CD13; gp150; h AP-N; hAPN; membrane alanyl aminopeptidase; Microsomal aminopeptidase; Myeloid plasma membrane glycoprotein CD13
Gene Aliases: ANPEP; APN; CD13; GP150; LAP1; P150; PEPN
UniProt ID: (Human) P15144
Entrez Gene ID: (Human) 290
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support