Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: IKYRKNEVFL DVIESVNLLV SANGNVLRSE IVGSIKMRVF LSGMPELRLG LNDKVLFDNT GRGKSKSVEL EDVKFHQCVR LSRFENDRTI SFIPPDGEFE LMSYRLNTHV KPLIWIESVI EKHSHSRIEY MIKAKSQFKR RSTANNVEIH IPVPNDADSP KFKTTVGSVK WVPENSEIVW SIKSFPGGKE YLMRAHFGLP SVEAEDKEGK PPISVKFEIP YFTTSGIQVR YLKIIEKSGY QALPWVRYIT QNGDYQLRTQ
The protein encoded by this gene is the medium chain of the trans-Golgi network clathrin-associated protein complex AP-1. The other components of this complex are beta-prime-adaptin, gamma-adaptin, and the small chain AP1S1. This complex is located at the Golgi vesicle and links clathrin to receptors in coated vesicles. These vesicles are involved in endocytosis and Golgi processing. Alternatively spliced transcript variants encoding distinct protein isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adapter-related protein complex 1 subunit mu-1; adaptor protein complex AP-1 mu-1 subunit; Adaptor protein complex AP-1 subunit mu-1; adaptor-related protein complex 1 mu 1 subunit; adaptor-related protein complex 1 mu-1 subunit; Adaptor-related protein complex 1 subunit mu-1; adaptor-related protein complex 1, mu 1 subunit; AP-1 complex subunit mu-1; AP-mu chain family member mu1A; clathrin adaptor protein AP47; clathrin assembly protein complex 1 medium chain 1; Clathrin assembly protein complex 1 mu-1 medium chain 1; clathrin assembly protein complex 1, medium chain; clathrin assembly protein complex AP1, mu subunit; Clathrin coat assembly protein AP47; Clathrin coat-associated protein AP47; golgi adaptor AP-1 47 kDa protein; Golgi adaptor HA1/AP1 adaptin mu-1 subunit; HA1 47 kDa subunit; Mu-adaptin 1; Mu1A-adaptin
Gene Aliases: [m]1A; AA408894; Adtm1A; AP1M1; AP47; CLAPM2; CLTNM; MU-1A; mu1A
UniProt ID: (Human) Q9BXS5, (Mouse) P35585
Entrez Gene ID: (Human) 8907, (Mouse) 11767
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support