Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: PAKRHEGDMY NMQYSSQQQE MYNQYGGSYS GPDRRPIQGQ YPYPYSRERM QGPGQIQTHG IPPQMMGGPL QSSSSEGPQQ NMWAARNDMP YPYQNRQGPG GPTQAPPYPG MNRTDDMMVP DQRINHESQW PSHVSQRQP
This locus encodes an AT-rich DNA interacting domain-containing protein. The encoded protein is a component of the SWI/SNF chromatin remodeling complex and may play a role in cell-cycle activation. The protein encoded by this locus is similar to AT-rich interactive domain-containing protein 1A. These two proteins function as alternative, mutually exclusive ARID-subunits of the SWI/SNF complex. The associated complexes play opposing roles. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq, Feb 2012].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARID domain-containing protein 1B; AT rich interactive domain 1B (SWI1-like); AT-rich interactive domain-containing protein 1B; BAF250B; BRG1-associated factor 250b; BRG1-binding protein ELD/OSA1; BRG1-binding protein hELD/OSA1; ELD (eyelid)/OSA protein; hOsa2; Osa homolog 2; p250R
Gene Aliases: 6A3-5; ARID1B; BAF250B; BRIGHT; CSS1; DAN15; ELD/OSA1; KIAA1235; MRD12; OSA2; P250R
UniProt ID: (Human) Q8NFD5
Entrez Gene ID: (Human) 57492
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support