Sulfatases (EC 3.1.5.6), such as ARSK, hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules (Sardiello et al., 2005 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: arylsulfatase family, member K; Arylsulfatase K; ASK; Telethon sulfatase
Gene Aliases: 2810429K17Rik; 4833414G15Rik; ARSK; RGD1310182; TSULF; UNQ630/PRO1246
UniProt ID: (Human) Q6UWY0, (Mouse) Q9D2L1, (Rat) Q32KJ2
Entrez Gene ID: (Human) 153642, (Mouse) 77041, (Rat) 365619
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support