Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: KRMARKNCLV KNLEAVETLG STSTICSDKT GTLTQNRMTV AHMWFDNQIH EADTTEDQSG ATFDKRSPTW TALSRIAGLC NRAVFKAGQE NISVSKRDTA GDASESALLK CIELSCGSVR KMRDRNPKVA EIPFNSTNKY QLSIHEREDS PQSHVLVMKG APERILDRCS TILVQGKEIP LDKEMQDAFQ NAYMELGGLG ERVLGFCQLN LPSGKFPRGF KFDTDELNFP TEKLCFVGLM SMIDPPRAAV PDAVGKCRSA GIKVIMVTGD HPITAKAIAK GVGIISEGNE TVEDIAARLN IPMSQVNPRE AKACVVHGSD LKDMTSEQLD EILKNHTEIV FARTSPQQKL IIVE
ATP1A1/2 is the catalytic component of the active enzyme Na+/K+-ATPase, which catalyzes the hydrolysis of ATP coupled with the exchange of sodium and potassium ions across the plasma membrane. The Na+/K+-ATPase is composed of a larger catalytic alpha-subunit (~110 kDa) and a small beta-subunit (~55 kDa). The alpha subunit has four isoforms identified to date: alpha 1, alpha 2, alpha 3 and alpha 4.The alpha 1 isoform is expressed ubiquitously but the alpha 2 isoform is present largely in the skeletal muscle, heart and vascular smooth muscle. The alpha 3 isoform is found almost exclusively in neurons and ovaries. The alpha 4 isoform is expressed in sperm.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (Na+ and K+) ATPase, alpha catalytic subunit precursor; (Na+ and K+) ATPase, alpha+ catalytic subunit precursor; (Na+, K+)-ATPase alpha-subunit; ATPase Na+/K+ transporting alpha 2 polypeptide; ATPase, Na+/K+ transporting, alpha 1 polypeptide; ATPase, Na+/K+ transporting, alpha 2 polypeptide; ATPase, Na+K+ transporting, alpha 1; ATPase, Na+K+ transporting, alpha 2; G00-119-711; Na(+)/K(+) ATPase alpha(+) subunit; Na(+)/K(+) ATPase alpha-1 subunit; Na(+)/K(+) ATPase alpha-2 subunit; Na+ ,K+ -ATPase alpha subunit; Na+ /K+ ATPase alpha 1 subunit; Na+ K+ ATPase alpha subunit; Na+, K+-ATPase beta-subunit precursor; Na+/K+ -ATPase alpha 2 subunit; Na+/K+ ATPase 1; Na+/K+ ATPase, alpha-A(+) catalytic polypeptide; Na+/K+ ATPase, alpha-B polypeptide; Na+/K+ transporting alpha 1 polypeptide; Na+/K+ transporting alpha 2 polypeptide; Na, K-ATPase, alpha-A catalytic polypeptide; Na,K-ATPase alpha 2 subunit; Na,K-ATPase alpha-1 subunit; Na,K-ATPase catalytic subunit alpha-A protein; Na,K-ATPase subunit alpha-B; Na,K-ATPase, alpha-subunit; sodium pump subunit alpha-1; Sodium pump subunit alpha-2; sodium-potassium ATPase catalytic subunit alpha-1; sodium-potassium ATPase catalytic subunit alpha-2; sodium-potassium-ATPase, alpha 1 polypeptide; sodium/potassium-transporting ATPase alpha-2 chain; unnamed protein product
Gene Aliases: Atpa-1; Atpa-3; CMT2DD; DEE98; FARIMPD; FHM2; HOMGSMR2; MHP2; mKIAA0778; Nkaa1b; RATATPA2
UniProt ID: (Human) P05023, (Pig) P05024, (Human) P50993, (Pig) D2WKD8, (Mouse) Q8VDN2, (Rat) P06685, (Mouse) Q6PIE5, (Rat) P06686
Entrez Gene ID: (Human) 476, (Pig) 397481, (Human) 477, (Pig) 396828, (Mouse) 11928, (Rat) 24211, (Mouse) 98660, (Rat) 24212
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support