Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody does not react with drosophila.
Suggested positive control: HEK293 cell lysate.
The Drosophila gene for the ATP synthase epsilon subunit (ATP5E) is known as stunted or sun (Kidd et al. 2005). Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP synthase epsilon chain, mitochondrial; ATP synthase F1 subunit epsilon; ATP synthase subunit epsilon, mitochondrial; ATP synthase, H transporting, mitochondrial F1 complex, epsilon subunit; ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit L homeolog; ATP synthase, H+ transporting, mitochondrial F1 complex, epsilon subunit; ATPase subunit epsilon; F(0)F(1)-ATPase; H(+)-transporting two-sector ATPase; mitochondrial ATP synthase epsilon chain; mitochondrial ATPase
Gene Aliases: ATP5E; ATP5F1E
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support