Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody is predicted to react with Monkey, Rabbit, Cat.
Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
Vasopressin receptor 1a (AVPR1A) is a receptor for arginine vasopressin. The activity of this receptor is mediated by G-proteins which activate a phosphatidyl-inositol-calcium second messenger system. AVPR1A has been involved in social behaviors, including affiliation and attachment.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antidiuretic hormone receptor 1a; AVPR V1a; SCCL vasopressin subtype 1a receptor; V1-vascular vasopressin receptor AVPR1A; V1a receptor; V1a vasopressin receptor; V1aR; Vascular/hepatic-type arginine vasopressin receptor; Vasopressin V1a receptor; Vasopressin V1aR
Gene Aliases: AVPR V1a; AVPR1; AVPR1A; V1aR
UniProt ID: (Human) P37288
Entrez Gene ID: (Human) 552
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support