Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • AXUD1 Antibodies

          Abnova

          AXUD1 Monoclonal Antibody (5E8)

          View all (8) AXUD1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite AXUD1 Monoclonal Antibody (5E8)

          Product Details

          H00064651-M01

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          ELISA (ELISA)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          5E8

          Immunogen

          AXUD1 (AAH38949, 1 a.a. approximately 589 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Immunogen sequence: MTGLLKRKFD QLDEDNSSVS SSSSSSGCQS RSCSPSSSVS RAWDSEEEGP WDQMPLPDRD FCGPRSFTPL SILKRARRER PGRVAFDGIT VFYFPRCQGF TSVPSRGGCT LGMALRHSAC RRFSLAEFAQ EQARARHEKL RQRLKEEKLE MLQWKLSAAG VPQAEAGLPP VVDAIDDASV EEDLAVAVAG GRLEEVSFLQ PYPARRRRAL LRASGVRRID REEKRELQAL RQSREDCGCH CDRICDPETC SCSLAGIKCQ MDHTAFPCGC CREGCENPMG RVEFNQARVQ THFIHTLTRL QLEQEAESFR ELEAPAQGSP PSPGEEALVP TFPLAKPPMN NELGDNSCSS DMTDSSTASS SASGTSEAPD CPTHPGLPGP GFQPGVDDDS LARILSFSDS DFGGEEEEEE EGSVGNLDNL SCFHPADIFG TSDPGGLASW THSYSGCSFT SGILDENANL DASCFLNGGL EGSREGSLPG TSVPPSMDAG RSSSVDLSLS SCDSFELLQA LPDYSLGPHY TSQKVSDSLD NIEAPHFPLP GLSPPGDASS CFLESLMGFS EPAAEALDPF IDSQFEDTVP ASLMEPVPV

          Target Information

          Axin, an important regulator of beta-catenin, is frequently mutated in human hepatocellular carcinomas (HCCs), and transduction of the wild-type Axin gene (AXIN1) induces apoptosis in HCC cells as well as in colon cancer cells. A novel gene, AXUD1 (AXIN1 up-regulated), has enhanced expression in response to exogenously expressed AXIN1. It is expressed in all human tissues examined, most abundantly in lung, placenta, skeletal muscle, pancreas and leukocytes. Data suggests that AXUD1 may have a tumor-suppressor function in those organs.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Axin-1 up-regulated gene 1 protein; AXIN1 up-regulated 1; CSRNP-1; Cysteine/serine-rich nuclear protein 1; DKFZp566F164; Protein URAX1; TAIP-3; TGF-beta-induced apoptosis protein 3; URAX1 protein

          View more View less

          Gene Aliases: AXUD1; CSRNP-1; CSRNP1; FAM130B; TAIP-3; TAIP3; URAX1

          View more View less

          UniProt ID: (Human) Q96S65

          View more View less

          Entrez Gene ID: (Human) 64651

          View more View less

          Function(s)
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding molecular_function transcription factor activity, sequence-specific DNA binding protein binding sequence-specific DNA binding
          Process(es)
          transcription from RNA polymerase II promoter apoptotic process post-embryonic development positive regulation of transcription from RNA polymerase II promoter platelet-derived growth factor receptor signaling pathway skeletal system morphogenesis palate development face morphogenesis
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-56756dfb8b-qtn7p:80/100.66.78.40:80.
          git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.1-2025.06.25-1.0