Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: VFWCTADIG WITGHSYVTY GPLANGATSV LFEGIPTYPD VNRLWSIVDK YKVTKFYTAP TAIRLLMKFG DEPVTKHSRA SLQVLGTVGE PINPEAWLWY HRVVGAQRCP IVDTFWQTET GGHMLTPLPG ATPMKPGSAT FPFFGVAPAI LNESGEELEG EAEGYLVFKQ PWPGIMRTVY GNHERFETTY FKKFPGYYVT GDGCQRDQDG YYWITGRIDD MLNVSGHLLS TAEVESALVE HEAVAEAAVV GHPHPVKGEC LYCFVTLCDG HTFSPKLTEE LKKQIREKIG PIATPDYIQN APGLPKTRSG KIMRRVLRKI AQNDHDLGDM STVADPSVIS HLFSHRCLTI Q (185-534 aa encoded by BC010141)
This gene encodes a protein with a sterol sensing domain (SSD) and seven WD domains. In the presence of cholesterol, this protein binds to sterol regulatory element binding proteins (SREBPs) and mediates their transport from the ER to the Golgi. The SREBPs are then proteolytically cleaved and regulate sterol biosynthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetate thiokinase; acetate--CoA ligase; acetate-CoA ligase; acetyl-CoA synthetase 1; acetyl-Coenzyme A synthetase 1 (AMP forming); acetyl-Coenzyme A synthetase 2 (ADP forming); acetyl-Coenzyme A synthetase 2 (AMP forming); ACS; acyl-activating enzyme; cytoplasmic acetyl-coenzyme A synthetase; lactyl-CoA synthetase; propionate--CoA ligase; propionyl-CoA synthetase; unnamed protein product
Gene Aliases: 1110017C11Rik; ACAS; Acas1; ACAS2; ACECS; AceCS1; ACS; Acs1; ACSA; dJ1161H23.1
UniProt ID: (Human) Q9NR19, (Mouse) Q9QXG4
Entrez Gene ID: (Human) 55902, (Rat) 311569, (Mouse) 60525
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support