Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. In vertebrates there are three groups of actin isoforms: alpha, beta and gamma. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exists in most cell types as components of the cytoskeleton and as mediators of internal cell motility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; Actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; Actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; actin, beta; Actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed; Actin, cytoplasmic 2; Actin, cytoplasmic 2, N-terminally processed; Actin, gamma-enteric smooth muscle; Actin, gamma-enteric smooth muscle, intermediate form; actin-like protein; Alpha-actin-1; Alpha-actin-2; Alpha-actin-3; Alpha-cardiac actin; alpha-sma; Atcb; beta cytoskeletal actin; Beta-actin; Cell growth-inhibiting gene 46 protein; cytoskeletal gamma-actin; deafness, autosomal dominant 20; deafness, autosomal dominant 26; epididymis luminal protein 176; Gamma-2-actin; Gamma-actin; nemaline myopathy type 3; pan actin; PS1TP5-binding protein 1; RP5-1068B5.2; Smooth muscle gamma-actin
Gene Aliases: AAT6; ACT; ACTA; ACTA1; ACTA2; ACTA3; ACTB; ACTC; ACTC1; ACTE; ACTG; ACTG1; ACTG2; ACTL3; ACTSA; ACTSG; ACTVS; ASD5; ASMA; BRWS1; BRWS2; CFTD; CFTD1; CFTDM; CMD1R; CMH11; DFNA20; DFNA26; GIG46; HEL-176; LVNC4; MPFD; MYMY5; NEM1; NEM2; NEM3; PS1TP5BP1; SHPM; VSCM
UniProt ID: (Human) P68133, (Human) P62736, (Human) P60709, (Human) P68032, (Human) P63261, (Human) P63267
Entrez Gene ID: (Human) 58, (Human) 59, (Human) 60, (Human) 70, (Human) 71, (Human) 72
Molecular Function: actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support