Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 7
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. In vertebrates there are three groups of actin isoforms: alpha, beta and gamma. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exists in most cell types as components of the cytoskeleton and as mediators of internal cell motility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A-X actin; A-X actin-like protein; ACTA1; ACTA2; ACTC; ACTG2; actin; actin alpha 1; actin alpha cardiac; Actin is a protein with a high degree of evolutionary conservation that undergoes polymerization to generate filaments, which subsequently assemble into interconnected networks within the cytoplasm of cells.; actin, alpha 2, smooth muscle, aorta; actin, alpha, cardiac 1; actin, alpha, vascular smooth muscle; actin, beta, cytoplasmic; actin, gamma 2, smooth muscle, enteric; actin, gamma, cytoplasmic 1; actin-like protein; alfa sma; alpha smooth muscle actin; alpha-actin; alpha-actin cardiac; alpha-actin-1; alpha-actin-2; alpha-actin-3; alpha-cardiac actin; alpha-sma; alpha-smooth muscle actin; alphac-actin; aSMA; aSmooth Muscle Actin; Atcb; beta cytoskeletal actin; beta-actin; beta-actin FE-3; cell growth-inhibiting gene 46 protein; cytoplasmic beta-actin; cytoplasmic protein; cytoskeletal gamma-actin; epididymis luminal protein 176; gamma-2-actin; gamma-actin; gamma-enteric smooth muscle actin; GIG46; I(2)-actin; melanoma X-actin; nemaline myopathy type 3; pan actin; PS1TP5-binding protein 1; skeletal muscle alpha-actin; sma; smooth muscle alpha-actin; smooth muscle gamma-actin; unnamed protein product
Gene Aliases: 0610041G09Rik; a-SMA; ACT; Act-4; Act4; ACTA; Acta-2; ACTA3; ACTC; Actc-1; ACTE; ACTG; ACTGE; Actl; ACTL3; Acts; ACTSA; ACTSG; Actsk-1; Actvs; Actx; alphaSMA; ASD5; ASMA; beta-actin; BKRNS; BNS; BRWS1; CFTD; CFTD1; CFTDM; CMD1R; CMH11; CMYO2A; CMYO2B; CMYO2C; CMYP2A; CMYP2B; CMYP2C; CSMH; DDS1; DFNA20; DFNA26; E430023M04Rik; E51; HEL-176; LVNC4; MMIHS5; MPFD; NEM1; NEM2; NEM3; PS1TP5BP1; SHPM; SMAalpha; SMalphaA; SMDYS; SMGA; THC8; VSCM; VSCM1; wu:fb63d03
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support