Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:1,000 | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:100 | - |
Immunocytochemistry (ICC/IF) |
1:100-1:500 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
A synthesized peptide derived from human ADCY5/6, corresponding to a region within the internal amino acids. |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS, pH 7.4, with 50% glycerol, 150mM NaCl |
Contains |
0.02% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2850723 |
Adenylate cyclase 5/6 encodes a member of the membrane-bound adenylyl cyclase enzymes. Adenylyl cyclases mediate G protein-coupled receptor signaling through the synthesis of the second messenger cAMP. Activity of the encoded protein is stimulated by the Gs alpha subunit of G protein-coupled receptors and is inhibited by protein kinase A, calcium and Gi alpha subunits. Single nucleotide polymorphisms in this gene may be associated with low birth weight and type 2 diabetes. Alternatively spliced transcript variants that encode different isoforms have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A cyclase V; A cyclase VI; AC5; AC6; ACVI; adcy5; Adenylate cyclase type 5; Adenylate cyclase type 6; Adenylate cyclase type V; Adenylate cyclase type VI; Adenylyl cyclase 5; Adenylyl cyclase 6; adenylyl cyclase type V; ATP pyrophosphate-lyase 5; ATP pyrophosphate-lyase 6; Ca(2+)-inhibitable adenylyl cyclase
Gene Aliases: AC5; AC6; ACVI; ADCY5; ADCY6; ADCYB; AW121902; FDFM; KIAA0422; LCCS8; mKIAA0422
UniProt ID: (Human) O95622, (Human) O43306, (Rat) Q04400, (Mouse) P84309, (Rat) Q03343, (Mouse) Q01341
Entrez Gene ID: (Human) 111, (Human) 112, (Rat) 64532, (Mouse) 224129, (Rat) 25289, (Mouse) 11512
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support