Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
MIA1703 targets Alkaline Phosphatase in ELISA, IHC, IP, and RIA applications and shows reactivity with Bovine samples.
The MIA1703 immunogen is purified Bovine Calf Intestine ALP.
MIA1703 detects Alkaline Phosphatase which has a predicted molecular weight of approximately 57 kDa.
MIA1703 was formerly sold as a Seradyn product.
There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The intestinal alkaline phosphatase gene encodes a digestive brush-border enzyme. This enzyme is upregulated during small intestinal epithelial cell differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Akp2; EC 3.1.3.1; I Kasahara isozyme; intestinal alkaline phosphatase V; Regan isozyme
Gene Aliases: IAP
Entrez Gene ID: (Bovine) 280993
Molecular Function: phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support