Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: QGDAAQKTD TSHHDQDHPT FNKITPNLAE FAFSLYRQLA HQSNSTNIFF SPVSIATAFA MLSLGTKADT HDEILEGLNF NLTEIPEAQI HEGFQELLRT LNQPDSQLQL TTGNGLFLSE GLKLVDKFLE DVKKLYHSEA FTVNFGDTEE AKKQINDYVE KGTQGKIVDL VKELDRDTVF ALVNYIFFKG KWERPFEVKD TEEEDFHVDQ ATTVKVPMMK RLGMFNIQHC KKLSSWVLLM KYLGNATAIF FLPDEGKLQH LENELTHDII TKFLENEDRR SASLHLPKLS ITGTYDLKSV LGQLGITKVF SNGADLSGVT EEAPLKLSKA VHKAVLTIDE KGTEAAGAMF LEAIPMSIPP EVKFNKPFVF LMIEQNTKSP LFMGKVVNPT QK (28-418 aa encoded by BC015642)
Alpha-1-AT is synthesized in the liver and it acts as an inhibitor of proteases such as trypsin, elastase, chymotrypsin, collagenase, leucocytic proteases, plasmin, and thrombin, which may be released during inflammatory reactions in the lung. In the absence of alpha-1-AT, these enzymes are not inhibited and they may digest pulmonary parenchyma. Alpha-1-AT deficiency is associated with chronic obstructive lung disease (emphysema) and less frequently with hepatic cirrhosis in infants and respiratory distress of the newborn. Increase in alpha-1-AT occurs as an acute phase response to tissue necrosis and inflammation. Serum level of alpha-1-AT is elevated in rheumatoid arthritis, bacterial infections, vasculitis, and carcinomatosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a-1 AT; a1-antitrypsin; AAT; alpha 1 antitrypsin; alpha-1 antitrypsin; Alpha-1 protease inhibitor; Alpha-1 protease inhibitor 1; Alpha-1-antiproteinase; Alpha-1-antitrypsin; alpha-1-antitrypsin (protease inhibitor); Alpha-1-antitrypsin 1-1; alpha-1-antitrypsin 2; alpha-1-antitrypsin null; alpha-1-protease inhibitor; Alpha-1-proteinase inhibitor; Digestive Zymogen; MGC23330; MGC9222; protease inhibitor 1 (anti-elastase), alpha-1-antitrypsin; Protease Serine 1; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1; serine (or cysteine) proteinase inhibitor, clade A, member 1; serine (or cysteine) proteinase inhibitor, clade A, member 1a; Serine Protease 1; Serine protease inhibitor 1-1; Serine protease inhibitor A1a; serine protease inhibitor alpha 1; Serpin A1; Serpin A1a; serpin peptidase inhibitor clade A member 1; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1; Short peptide from AAT; SPAAT; TRP1; TRY1
Gene Aliases: A1A; A1AT; AAT; Aat-2; Aat2; alpha1AT; Dom1; PI; PI1; PRO0684; PRO2209; PRO2275; SERPINA1; Serpina1a; Spi1; Spi1-1; Spi1-3
UniProt ID: (Pig) P50447, (Human) B2RDQ8, (Rat) P17475, (Mouse) P07758
Entrez Gene ID: (Pig) 397688, (Human) 5265, (Rat) 24648, (Mouse) 20700
Molecular Function: protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support