Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • alpha Actinin 1 Antibodies

          Proteintech

          Alpha Actinin Monoclonal Antibody (1E2E1), CoraLite® 594

          View all (18) alpha Actinin 1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Alpha Actinin Monoclonal Antibody (1E2E1), CoraLite® 594

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          Alpha Actinin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Alpha Actinin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Alpha Actinin Antibody (CL594-66895) in ICC/IF

          Immunofluorescent analysis of (-20°C Methanol) fixed C2C12 cells using CoraLite®594 Alpha Actinin antibody (CL594-66895, Clone: 1E2E1) at dilution of 1:100. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Alpha Actinin Antibody in Immunocytochemistry (ICC/IF)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Alpha Actinin Monoclonal Antibody (1E2E1), CoraLite® 594

          Product Details

          CL594-66895

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1E2E1

          Immunogen

          Alpha Actinin fusion protein Ag28383
          3D Epitope / Immunogen

          Conjugate

          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation

          Excitation/Emission Max

          593/614 nm

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.5% BSA

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: EIQGLTTAHE QFKATLPDAD KERLAILGIH NEVSKIVQTY HVNMAGTNPY TTITPQEING KWDHVRQLVP RRDQALTEEH ARQQHNERLR KQFGAQANVI GPWIQTKMEE IGRISIEMHG TLEDQLSHLR QYEKSIVNYK PKIDQLEGDH QLIQEALIFD NKHTNYTMEH IRVGWEQLLT TIARTINEVE NQILTRDAKG ISQEQMNEFR ASFNHFDRDH SGTLGPEEFK ACLISLGYDI GNDPQGEAEF ARIMSIVDPN RLGVVTFQAF IDFMSRETAD TDTADQVMAS FKILAGDKNY ITMDELRREL PPDQAEYCIA RMAPYTGPDS VPGALDYMSF STALYGESDL

          Target Information

          Alpha Actinin 1 belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a nonmuscle, cytoskeletal, alpha actinin isoform and maps to the same site as the structurally similar erythroid beta spectrin gene. Three transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: actinin 1 smooth muscle; alpha actinin 1a; bs-alpha actinin 1; epididymis secretory sperm binding protein; F-actin cross-linking protein; FLJ40884; FLJ54432; non-muscle alpha-actinin-1; unnamed protein product

          View more View less

          Gene Aliases: 3110023F10Rik; Actn1a; BDPLT15

          View more View less

          UniProt ID: (Human) P12814, (Mouse) Q7TPR4, (Rat) Q9Z1P2

          View more View less

          Entrez Gene ID: (Human) 87, (Mouse) 109711, (Rat) 81634

          View more View less

          Function(s)
          transcription coactivator activity double-stranded RNA binding integrin binding calcium ion binding protein binding cytoskeletal protein binding vinculin binding identical protein binding protein homodimerization activity ion channel binding actin filament binding structural constituent of postsynapse cytoskeletal regulatory protein binding protein domain specific binding
          Process(es)
          actin filament organization actin cytoskeleton organization platelet formation cortical cytoskeleton organization platelet morphogenesis positive regulation of transcription, DNA-templated focal adhesion assembly actin filament bundle assembly actin filament network formation muscle cell development postsynapse organization platelet activation regulation of apoptotic process anatomical structure formation involved in morphogenesis platelet aggregation fibril organization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch