Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Immunohistochemistry (IHC) |
Assay-Dependent | View 7 publications 7 publications |
Immunohistochemistry (Paraffin) (IHC (P)) |
- | View 2 publications 2 publications |
Product Specifications | |
---|---|
Species Reactivity |
Rat |
Published species |
Rat |
Host/Isotype |
Mouse / IgG1, kappa |
Class |
Monoclonal |
Type |
Antibody |
Clone |
JG12 |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS |
Contains |
no preservative |
Storage conditions |
-20°C |
RRID |
AB_10597125 |
Description: Aminopeptidase P is a glycosyl-phosphatidylinositol (GPI) anchored membrane enzyme that is recognized by a monoclonal antibody designated JG12 that was raised against rat glomerular membrane protein fractions and labeled intensely microvessels of kidney and lung.
Applications Tested: Immunohistochemistry (Paraffin).
This gene encodes the cytosolic form of a metalloaminopeptidase that catalyzes the cleavage of the N-terminal amino acid adjacent to a proline residue. The gene product may play a role in degradation and maturation of tachykinins, neuropeptides, and peptide hormones. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aminoacylproline aminopeptidase; cytoplasmic aminopeptidase P; Cytosolic aminopeptidase P; OTTHUMP00000020457; OTTHUMP00000058856; sAmp; Soluble aminopeptidase P; X-Pro aminopeptidase 1; X-Pro aminopeptidase 1, soluble; X-prolyl aminopeptidase 1, soluble; Xaa-Pro aminopeptidase 1; xpnpep1 {ECO:0000312|RGD:621274}
Gene Aliases: App; Xpnpep1
UniProt ID: (Rat) O54975
Entrez Gene ID: (Rat) 170751
Molecular Function:
protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support