Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MSTVHEILC KLSLEGDHST PPSAYGSVKA YTNFDAERDA LNIETAIKTK GVDEVTIVNI LTNRSNAQRQ DIAFAYQRRT KKELASALKS ALSGHLETVI LGLLKTPAQY DASELKASMK GLGTDEDSLI EIICSRTNQE LQEINRVYKE MYKTDLEKDI ISDTSGDFRK LMVALAKGRR AEDGSVIDYE LIDQDARDLY DAGVKRKGTD VPKWISIMTE RSVPHLQKVF DRYKSYSPYD MLESIRKEVK GDLENAFLNL VQCIQNKPLY FADRLYDSMK GKGTRDKVLI RIMVSRSEVD MLKIRSEFKR KYGKSLYYYI QQDTKGDYQK ALLYLCGGDD (1-339 aa encoded by BC016774)
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions as an autocrine factor which heightens osteoclast formation and bone resorption. This gene has three pseudogenes located on chromosomes 4, 9 and 10, respectively. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 33-kDa calcimedin; 33-kDa lymphocyte Ca<2+>- binding protein; 33kDa lymphocyte Ca<2+>- binding protein; 36 kDa calcium-dependent phospholipid-binding protein; 36-kDa calelectrin; annexin II; annexin-2; calpactin I heavy chain; calpactin I heavy chain (p36); calpactin I heavy polypeptide; calpactin-1 heavy chain; chromobindin 8; chromobindin-8; epididymis secretory protein Li 270; epididymis secretory sperm binding protein; lipocortin II; p36; PAP-IV; placental anticoagulant protein IV; protein I; unnamed protein product
Gene Aliases: ANX2; ANX2L4; CAL1H; HEL-S-270; LIP2; LPC2; LPC2D; P36; PAP-IV
UniProt ID: (Human) P07355, (Rat) Q07936, (Mouse) P07356
Entrez Gene ID: (Human) 302, (Rat) 56611, (Mouse) 12306
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support