Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:2,000-1:16,000 | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:50-1:500 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Mouse / IgG2a |
Class |
Monoclonal |
Type |
Antibody |
Clone |
1A11E4 |
Immunogen |
Annexin A3 Fusion Protein Ag2381 (1-322 aa encoded by BC000871) |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein A |
Storage buffer |
PBS, pH 7.3, with 50% glycerol |
Contains |
0.02% sodium azide |
Storage conditions |
-20°C |
Immunogen sequence: MASIWVGHR GTVRDYPDFS PSVDAEAIQK AIRGIGTDEK MLISILTERS NAQRQLIVKE YQAAYGKELK DDLKGDLSGH FEHLMVALVT PPAVFDAKQL KKSMKGAGTN EDALIEILTT RTSRQMKDIS QAYYTVYKKS LGDDISSETS GDFRKALLTL ADGRRDESLK VDEHLAKQDA QILYKAGENR WGTDEDKFTE ILCLRSFPQL KLTFDEYRNI SQKDIVDSIK GELSGHFEDL LLAIVNCVRN TPAFLAERLH RALKGIGTDE FTLNRIMVSR SEIDLLDIRT EFKKHYGYSL YSAIKSDTSG DYEITLLKIC GGD (1-322 aa encoded by BC000871)
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions in the inhibition of phopholipase A2 and cleavage of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. This protein may also play a role in anti-coagulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 35-alpha calcimedin; Annexin 3; Annexin A3; Annexin III; Annexin III (lipocortin III); annexin III (lipocortin III, 1,2-cyclic-inositol-phosphate phosphodiesterase, placental anticoagulant protein III, calcimedin 35-alpha); Annexin-3; Annexin3; ANXA 3; calcimedin 35-alpha; Inositol 1,2-cyclic phosphate 2-phosphohydrolase; Lipocortin III; PAP-III; Placental anticoagulant protein III
Gene Aliases: ANX3; ANXA3
UniProt ID: (Human) P12429
Entrez Gene ID: (Human) 306
Molecular Function:
calcium-binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support