Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1 µg/mL | View 1 publication 1 publication |
Immunocytochemistry (ICC/IF) |
1-5 µg/mL | - |
ELISA (ELISA) |
0.5 µg/mL | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Published species |
Mouse |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Synthetic peptide conjugated to KLH derived from within residues 200 to the C-terminus of Human Apolipoprotein A I. |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
0.5 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
PBS, pH 7.4, with 1% BSA |
Contains |
0.02% sodium azide |
Storage conditions |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
RRID |
AB_10989874 |
This antibody is predicted to react with dog, pig and baboon based on sequence homology.
Apolipoprotein A-1 (apo A-1) in human plasma is a major protein component of high density lipoprotein (HDL). The molecular mass is 24 kDa. Apo A-1 is a cofactor for enzymes involved in the normal turnover of cholesterol and is produced in the liver. Plasma concentration is 270-380 mg/100 mL.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-Sep; 2-Sep; apo A-1; APO A1; APO I; Apo-AI; ApoAI; Apolipoprotein; Apolipoprotein A-I; Apolipoprotein A-I(1-242); Apolipoprotein A1; Apolipoprotein AI; MGC102525; MGC117399; MGC124929; ProapoA-I; Proapolipoprotein A-I; Truncated apolipoprotein A-I; Udpgt; Ugt1; Ugt1a7
Gene Aliases: apo(a); APOA1
UniProt ID: (Human) P02647
Entrez Gene ID: (Human) 335
Molecular Function:
apolipoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support