Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
By sandwich ELISA, MA5-14667 can be used as a detection antibody with Product # 710263 as the coating antibody, to generate a matched pair. Using this matched pair, recombinant human Apo A-1, but not recombinant mouse Apo A-1, was detected. It is not recommended to use Product # 701239 for coating when MA5-14667 is used for detection. By sandwich ELISA, MA5-14667 can be used to detect Apo A-1 from serum samples. To increase sensitivity of sandwich ELISAs with MA5-14667, a biotinylated detection antibody followed by Streptavidin-HRP is recommended.
By Western blot, MA5-14667 detects recombinant human Apo A-1, but not recombinant mouse Apo A-1. MA5-14667 may not be as successful for detecting endogenous Apo A-1 by Western blot.
Product MA514667 is a smaller package size of MIA1401 (formerly sold as a Seradyn product).
Apolipoprotein A-1 (apo A-1) in human plasma is a major protein component of high density lipoprotein (HDL). The molecular mass is 24 kDa. Apo A-1 is a cofactor for enzymes involved in the normal turnover of cholesterol and is produced in the liver. Plasma concentration is 270-380 mg/100 mL.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-Sep; 2-Sep; Apo a; apo A-1; APO A1; APO I; Apo-AI; ApoAI; Apolipoprotein; Apolipoprotein A-I; Apolipoprotein A1; Apolipoprotein AI; MGC102525; MGC117399; MGC124929; Udpgt; Ugt1; Ugt1a7
Gene Aliases: apo(a); APOA1
UniProt ID: (Human) P02647
Entrez Gene ID: (Human) 335
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support