Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Aquaporin 3 Antibodies

          Invitrogen

          Aquaporin 3 Recombinant Rabbit Monoclonal Antibody (PSH06-52)

          Advanced Verification
          View all (18) Aquaporin 3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Aquaporin 3 Recombinant Rabbit Monoclonal Antibody (PSH06-52)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (20)
          • Advanced Verification (6)
          Aquaporin 3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Aquaporin 3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 26

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Aquaporin 3 Antibody (MA562412) in ICC/IF

          Immunofluorescence analysis of Aquaporin 3 was performed using 70% confluent log phase HeLa cells. The cells were fixed with 4% paraformaldehyde for 10 minutes and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with Aquaporin 3 Recombinant Rabbit Monoclonal Antibody (PSH06-52)(Product # MA562412) at 1:50 dilution in 0.1% BSA, incubated at 4 degree Celsius overnight and then label... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Aquaporin 3 Antibody in Immunocytochemistry (ICC/IF)
          Aquaporin 3 Antibody in Immunocytochemistry (ICC/IF)
          Aquaporin 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aquaporin 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aquaporin 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aquaporin 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aquaporin 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aquaporin 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aquaporin 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aquaporin 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aquaporin 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aquaporin 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aquaporin 3 Antibody in Western Blot (WB)
          Aquaporin 3 Antibody in Western Blot (WB)
          Aquaporin 3 Antibody in Western Blot (WB)
          Aquaporin 3 Antibody in Western Blot (WB)
          Aquaporin 3 Antibody in Western Blot (WB)
          Aquaporin 3 Antibody in Multiplex Immunohistochemistry (mIHC)
          Aquaporin 3 Antibody in Multiplex Immunohistochemistry (mIHC)
          Aquaporin 3 Antibody in Multiplex Immunohistochemistry (mIHC)
          Aquaporin 3 Antibody
          Aquaporin 3 Antibody
          Aquaporin 3 Antibody
          Aquaporin 3 Antibody
          Aquaporin 3 Antibody
          Aquaporin 3 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Aquaporin 3 Recombinant Rabbit Monoclonal Antibody (PSH06-52)

          Product Details

          MA562412

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Multiplex Immunohistochemistry (mIHC)

          1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG1

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          PSH06-52

          Immunogen

          Synthetic peptide within human Aquaporin 3 aa 243-292.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.06 mg/mL

          Amount

          106 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4, with 0.1% BSA, 40% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3703598

          Product Specific Information

          Positive Control: HeLa cell lysates, human skin tissue, mouse skin tissue, rat skin tissue, HeLa.

          Tissue Specificity: Tissue enhanced (esophagus, skin, urinary bladder).

          Subcellular Location: Cell membrane, Basolateral cell membrane.

          Sequence Similarities: 96% Mouse/Rat.

          Predicted band size: 32 kDa.

          Target Information

          AQP3 is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 3 is localized at the basal lateral membranes of collecting duct cells in the kidney. In addition to its water channel function, aquaporin 3 has been found to facilitate the transport of nonionic small solutes such as urea and glycerol, but to a smaller degree. It has been suggested that water channels can be functionally heterogeneous and possess water and solute permeation mechanisms.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 31.4 kDa water channel protein; AQP 3; AQP-3; AQP3; aquaglyceroporin-3; aquaporin 3 (GIL blood group); Aquaporin3; Gill blood group; kidney collecting duct basolateral membrane water and glycerol permeable aquaporin; unnamed protein product

          View more View less

          Gene Aliases: AQP-2; AQP-3; GIL

          View more View less

          UniProt ID: (Human) Q92482, (Rat) P47862, (Mouse) Q8R2N1

          View more View less

          Entrez Gene ID: (Human) 360, (Rat) 65133, (Mouse) 11828

          View more View less

          Function(s)
          protein binding urea transmembrane transporter activity water channel activity glycerol channel activity channel activity identical protein binding polyol transmembrane transporter activity transmembrane transporter activity
          Process(es)
          positive regulation of immune system process renal water homeostasis water transport glycerol transport response to retinoic acid response to vitamin D odontogenesis regulation of keratinocyte differentiation response to calcium ion transmembrane transport cellular response to hypoxia urea transmembrane transport response to ischemia polyol transport urea transport renal water absorption
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-p95z8:80/100.66.74.151:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline