Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Recommended positive controls: Molt-4, Raji, U87-MG, SK-N-SH, IMR32, SK-N-AS.
Predicted reactivity: Mouse (87%), Rat (84%), Pig (95%), Rhesus Monkey (97%), Bovine (93%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ASPA is an enzyme that catalyzes the conversion of N-acetyl_L-aspartic acid (NAA) to aspartate and acetate. NAA is abundant in the brain where hydrolysis by aspartoacylase is thought to help maintain white matter. This protein is an NAA scavenger in other tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACY-2; aminoacylase 2; Aminoacylase-2; Aspartoacylase; aspartoacylase (aminoacylase) 2
Gene Aliases: Acy-2; ACY2; ASP; ASPA; nur7
UniProt ID: (Human) P45381, (Mouse) Q8R3P0
Entrez Gene ID: (Human) 443, (Mouse) 11484
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support