Search Thermo Fisher Scientific
MA1-26526 detects BCA225 from human samples.
MA1-26526 has been successfully used in immunohistochemistry (paraffin and frozen) procedures.
The MA1-26526 immunogen is BCA 225 protein secreted by the T47D (clone 11) human breast carcinoma cell line.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
BCA225 is a 225 kDa glycoprotein secreted by the T47D breast carcinoma cell line.
Protein Aliases: BCA 225