Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • BMPR1A Antibodies

          Alomone Labs, LTD

          BMPR1A (extracellular) Polyclonal Antibody

          View all (18) BMPR1A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite BMPR1A (extracellular) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          BMPR1A (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          BMPR1A (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          BMPR1A (extracellular) Antibody (ABR-043-200UL) in IHC (F)

          Expression of BMPR1A in mouse olfactory bulb. Immunohistochemical staining of perfusion-fixed frozen mouse brain sections with Anti-BMPR1A (extracellular) Antibody (#ABR-043) (1:300), followed by goat anti-rabbit-AlexaFluor-488. A. BMPR1A immunoreactivity (green), appears in cells lined around glomeruli (G, down pointing arrows) and in cells of the mitral layer (up pointing arrows). B. Pre-incubation of the... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          BMPR1A (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          BMPR1A (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          BMPR1A (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          BMPR1A (extracellular) Antibody in Western Blot (WB)
          BMPR1A (extracellular) Antibody in Western Blot (WB)
          BMPR1A (extracellular) Antibody in Western Blot (WB)
          BMPR1A (extracellular) Antibody in Flow Cytometry (Flow)
          BMPR1A (extracellular) Polyclonal Antibody

          Product Details

          ABR-043-200UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:300
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          Flow Cytometry (Flow)

          5 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Peptide APVHRDWRVHLALD, corresponding to amino acid residues 291-304 of mouse GPR142. Extracellular, 3rd loop.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.8 mg/mL

          Amount

          160 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 1% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Immunogen sequence: APVHRDWRVHLALD

          Reconstitution: 50 uL double distilled water (DDW). The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).

          Target Information

          BMPR1A is a Type I member of the TGF beta receptor superfamily of transmembrane ser/thr kinases which phosphorylates intracellular SMADs in response to bone morphogenic proteins. These receptors are also closely related to the activin receptors, ACVR1 and ACVR2. The ligands of these receptors are members of the TGF-beta superfamily. TGF-betas and activins transduce their signals through the formation of heteromeric complexes with 2 different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kD and type II receptors of about 70-80 kD. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: activin A receptor, type II-like kinase 3; Activin receptor-like kinase 3; ALK-3; BMP type-1A receptor; BMP-2/BMP-4 receptor; BMPR-1A; BMPRIA; Bone morphogenetic protein 4 receptor; Bone morphogenetic protein receptor type-1A; bone morphogenetic protein receptor, type 1A; bone morphogenetic protein receptor, type IA; CD292; Serine/threonine-protein kinase receptor R5; SKR5; unnamed protein product

          View more View less

          Gene Aliases: 10q23del; 1110037I22Rik; ACVRLK3; ALK-3; ALK3; AU045487; Bmpr; BMPR-1A; BMPR-IA; BMPR1A; CD292; SKR5

          View more View less

          UniProt ID: (Human) P36894, (Rat) Q78EA7, (Mouse) P36895

          View more View less

          Entrez Gene ID: (Human) 657, (Rat) 81507, (Mouse) 12166

          View more View less

          Function(s)
          nucleotide binding protein kinase activity protein serine/threonine kinase activity transmembrane receptor protein serine/threonine kinase activity transforming growth factor beta receptor activity, type I protein binding ATP binding kinase activity transferase activity BMP binding protein homodimerization activity SMAD binding metal ion binding BMP receptor activity RNA polymerase II transcription factor activity, sequence-specific DNA binding molecular_function receptor signaling protein serine/threonine kinase activity transforming growth factor beta-activated receptor activity glycoprotein binding transferase activity, transferring phosphorus-containing groups serine/threonine protein kinase receptor
          Process(es)
          angiogenesis osteoblast differentiation in utero embryonic development mesoderm formation somitogenesis Mullerian duct regression positive regulation of mesenchymal cell proliferation chondrocyte differentiation heart morphogenesis outflow tract septum morphogenesis outflow tract morphogenesis cardiac conduction system development atrioventricular valve development mitral valve morphogenesis tricuspid valve morphogenesis endocardial cushion morphogenesis cardiac right ventricle morphogenesis ventricular trabecula myocardium morphogenesis ventricular compact myocardium morphogenesis endocardial cushion formation cellular iron ion homeostasis immune response transmembrane receptor protein serine/threonine kinase signaling pathway transforming growth factor beta receptor signaling pathway pattern specification process ectoderm development nervous system development endoderm development heart development dorsal/ventral axis specification anterior/posterior pattern specification dorsal/ventral pattern formation positive regulation of gene expression negative regulation of gene expression regulation of cardiac muscle cell apoptotic process neural crest cell development negative regulation of smooth muscle cell migration central nervous system neuron differentiation pituitary gland development neural plate mediolateral regionalization cell differentiation lung development positive regulation of bone mineralization BMP signaling pathway positive regulation of transforming growth factor beta2 production somatic stem cell population maintenance hindlimb morphogenesis dorsal aorta morphogenesis odontogenesis of dentin-containing tooth embryonic digit morphogenesis positive regulation of osteoblast differentiation positive regulation of transcription from RNA polymerase II promoter paraxial mesoderm development paraxial mesoderm structural organization lateral mesoderm development regulation of lateral mesodermal cell fate specification mesendoderm development embryonic organ development developmental growth embryonic morphogenesis stem cell differentiation epithelial cell proliferation positive regulation of epithelial cell proliferation negative regulation of neurogenesis negative regulation of muscle cell differentiation cartilage development palate development regulation of cardiac muscle cell proliferation positive regulation of cardiac muscle cell proliferation positive regulation of SMAD protein import into nucleus ventricular septum morphogenesis neural plate pattern specification heart formation atrioventricular node cell development pharyngeal arch artery morphogenesis cellular response to growth factor stimulus cellular response to BMP stimulus positive regulation of dendrite development positive regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of cardiac ventricle development positive regulation of vascular smooth muscle cell proliferation fibrous ring of heart morphogenesis regulation of neural crest cell differentiation anti-Mullerian hormone signaling pathway regulation of cellular senescence regulation of endothelial cell proliferation transcription from RNA polymerase II promoter protein phosphorylation biological_process positive regulation of pathway-restricted SMAD protein phosphorylation stem cell population maintenance signal transduction by protein phosphorylation regulation of endothelial cell differentiation positive regulation of transcription, DNA-templated phosphorylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-548c9458fd-kpf9c:80/100.66.77.199:80.
          git-commit: b50e517baec6e51dce58f17cb59caa961a96861c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.53.0-Offline