Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
May be a bridge molecule between BCL2 and ARHGAP1/CDC42 in promoting cell death.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bcl-2/adenovirus E1B 19 kDa-interacting protein 2-like protein; BCL2/adenovirus E1B 19kD interacting protein like; BNIP-2 similar
Gene Aliases: BNIP-S; BNIP-Salpha; BNIP-Sbeta; BNIPL; BNIPL-1; BNIPL-2; BNIPL1; BNIPL2; BNIPS; PP753
UniProt ID: (Human) Q7Z465
Entrez Gene ID: (Human) 149428
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support