The recommended shelf life for this product is 1 year from date of receipt.
CDC42, a small Rho GTPase, regulates the formation of F-actin-containing structures through its interaction with the downstream effector proteins. The protein encoded by this gene is a member of the Borg family of CDC42 effector proteins. Borg family proteins contain a CRIB (Cdc42/Rac interactive-binding) domain. They bind to, and negatively regulate the function of, CDC42. This protein can interact with CDC42, as well as with the ras homolog gene family, member Q (ARHQ/TC10). Expression of this protein in fibroblasts has been shown to induce pseudopodia formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Binder of Rho GTPases 2; CDC42 effector protein (Rho GTPase binding) 3; Cdc42 effector protein 3; CRIB-containing BORG2 protein; MSE55-related Cdc42-binding protein; MSE55-related protein
Gene Aliases: BORG2; CDC42EP3; CEP3; UB1
UniProt ID: (Human) Q9UKI2
Entrez Gene ID: (Human) 10602
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support