Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:2,000 | View 1 publication 1 publication |
Immunohistochemistry (IHC) |
1:1,000 | - |
ChIP assay (ChIP) |
5-10 µL | View 1 publication 1 publication |
Miscellaneous PubMed (Misc) |
- | View 1 publication 1 publication |
ChIP-sequencing (ChIP-Seq) |
- | View 2 publications 2 publications |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse |
Published species |
Human, Mouse, Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
This BRD4 antibody was raised against a peptide within the C-terminal region of human BRD4. |
Conjugate |
Unconjugated |
Form |
Liquid |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; the longer variant has a C-terminus region with a praline and glutamine-rich domains. BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of the cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BRD4-NUT FUSION; bromodomain-containing 4; bromodomain-containing 5; Bromodomain-containing protein 4; chromosome-associated protein; fsh-like protein; MCAP; Mitotic chromosome-associated protein; Protein HUNK1
Gene Aliases: BRD4; HUNK1; Mcap
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support