Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is predicted to react with human based on sequence homology.
Store antibody at 4°C for 1-2 weeks. For long-term storage, store at -20°C.
BRN3A is a class IV POU domain-containing transcription factor highly expressed in the developing sensory nervous system and in cells of the B- and T-lymphocytic lineages.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain-3A; Brain-specific homeobox/POU domain protein 3A; Brn-3.0; brn-3A; FLJ13449; POU domain, class 4, transcription factor 1
Gene Aliases: Brn-3; Brn3; Brn3a; Pou4f1
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support