Product References
Estrogen Receptor-β Up-Regulates IGF1R Expression and Activity to Inhibit Apoptosis and Increase Growth of Medulloblastoma.
Endocrinology
Cookman CJ,Belcher SM
MA5-13666 was used in immunohistochemistry - paraffin section and western blot to determine the contribution of ESR2 to medulloblastoma growth in vitro and in vivo.
Wed Jul 01 00:00:00 EDT 2015
Estrogen Receptor-β Up-Regulates IGF1R Expression and Activity to Inhibit Apoptosis and Increase Growth of Medulloblastoma.
Endocrinology
Cookman CJ,Belcher SM
MA5-13666 was used in immunohistochemistry - paraffin section and western blot to determine the contribution of ESR2 to medulloblastoma growth in vitro and in vivo.
Wed Jul 01 00:00:00 EDT 2015
Effects of estrogen replacement therapy on apoptosis and vascular endothelial growth factor expression in ocular surface epithelial cells: An experimental study.
International journal of ophthalmology
Ozcura F,Dündar SO,Cetin ED,Beder N,Dündar M
MA5-13666 was used in immunohistochemistry to study rat ocular surface apoptosis and VEGF expression following estrogen replacement therapy
Thu Aug 23 00:00:00 EDT 2012
Immunohistochemistry with apoptotic-antiapoptotic proteins (p53, p21, bax, bcl-2), c-kit, telomerase, and metallothionein as a diagnostic aid in benign, borderline, and malignant serous and mucinous ovarian tumors.
Diagnostic pathology
Ozer H,Yenicesu G,Arici S,Cetin M,Tuncer E,Cetin A
MA5-13666 was used in immunohistochemistry to study the diagnostic value of the immunohistochemical expression of apoptotic and anti-apoptotic markers in serous and mucinous ovarian tumors
Thu Sep 20 00:00:00 EDT 2012
Clinicopathological significance of cathepsin D expression in non-small cell lung cancer is conditional on apoptosis-associated protein phenotype: an immunohistochemistry study.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Fan C,Lin X,Wang E
MA5-13666 was used in immunohistochemistry to study the influence of the phenotype of apoptosis-associated proteins on the clinical significance of cathepsin D expression in non-small cell lung cancer
Wed Aug 01 00:00:00 EDT 2012
The expression of IgM is helpful in the differentiation of primary cutaneous diffuse large B cell lymphoma and follicle center lymphoma.
Leukemia research
Demirkesen C,Tüzüner N,Esen T,Lebe B,Ozkal S
MA5-13666 was used in immunohistochemistry to investigate the effect of IgM expressoin on the differentiation of primary cutaneous diffuse large B cell lymphoma and follicle center lymphoma
Thu Sep 01 00:00:00 EDT 2011
Recurrence of benign meningiomas: predictive value of proliferative index, BCL2, p53, hormonal receptors and HER2 expression.
British journal of neurosurgery
Abdelzaher E,El-Gendi SM,Yehya A,Gowil AG
MA5-13666 was used in immunohistochemistry to identify biomarkers that predict the recurrence of benign meningiomas
Thu Dec 01 00:00:00 EST 2011
Radiotherapy in laryngeal carcinoma: can a panel of 13 markers predict response?
The Laryngoscope
Wildeman MA,Gibcus JH,Hauptmann M,Begg AC,van Velthuysen ML,Hoebers FJ,Mastik MF,Schuuring E,van der Wal JE,van den Brekel MW
MA5-13666 was used in immunohistochemistry to study the value of 13 immunohistochemical biomarkers in predicting response to radiotherapy in laryngeal cancer
Sun Feb 01 00:00:00 EST 2009
Curcumin-induced apoptosis in human leukemia cell HL-60 is associated with inhibition of telomerase activity.
Molecular and cellular biochemistry
Mukherjee Nee Chakraborty S,Ghosh U,Bhattacharyya NP,Bhattacharya RK,Dey S,Roy M
MA5-13666 was used in western blot to study the potential role of telomerase inhibition in the pro-apoptotic effects of curcumin in a human leukemic cell line
Thu Mar 01 00:00:00 EST 2007
Prognostic significance of cell proliferation and apoptosis-regulating proteins in Epstein-Barr virus positive and negative pediatric Hodgkin lymphoma.
Lymphatic research and biology
Aktaş S,Kargi A,Olgun N,Diniz G,Erbay A,Vergin C
MA5-13666 was used in immunohistochemistry to evaluate prognostic markers for Epstein-Barr virus positive and negative pediatric Hodgkin lymphoma
Mon Mar 03 00:00:00 EST 2008
Incidental calcifying fibrous tumor of the stomach presenting as a polyp.
Pathology international
Elpek GO,Küpesiz GY,Ogüs M
MA5-13666 was used in immunohistochemistry to report on a case of incidental calcifying fibrous tumor of the stomach presenting as a polyp
Sat Apr 01 00:00:00 EST 2006
Tea-induced apoptosis in human leukemia K562 cells as assessed by comet formation.
Asian Pacific journal of cancer prevention : APJCP
Chakraborty S,Kundu T,Dey S,Bhattacharya RK,Siddiqi M,Roy M
MA5-13666 was used in western blot to study tea-induced apoptosis in human leukemia K562 cells
Tue Sep 12 00:00:00 EDT 2006
Induction of apoptosis in human leukemia cells by black tea and its polyphenol theaflavin.
Cancer letters
Kundu T,Dey S,Roy M,Siddiqi M,Bhattacharya RK
MA5-13666 was used in western blot to study the mechanism of apoptosis induction in leukemic cells by the black tea polyphenol theaflavin
Thu Dec 08 00:00:00 EST 2005
Prognostic factors in transitional cell cancer of the bladder: an emerging role for Bcl-2 and p53.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology
Ong F,Moonen LM,Gallee MP,ten Bosch C,Zerp SF,Hart AA,Bartelink H,Verheij M
MA5-13666 was used in immunohistochemistry to study the prognostic value of Bcl-2 and p53 in transitional cell cancer of the bladder
Thu Nov 01 00:00:00 EST 2001