Search Thermo Fisher Scientific
MA5-14691 targets Benzoylecgonine in ELISA, IP, and RIA applications and shows reactivity with Chemical samples.
The MA5-14691 immunogen is benzoylecgonine-KLH.
Product MA514691 is a smaller package size of MIB0202 (formerly sold as a Seradyn product).
Benzoylecgonine is used as the main pharmaceutical ingredient in the prescription drug Esterom, a topical solution used for the relief of muscle pain.
Protein Aliases: C16H19NO4