Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Despite the predicted molecular weight, Bfl-1 often migrates at a higher molecular weight in SDS-PAGE, presumably due to post-translational modifications. A suggested positive control is mouse kidney tissue lysate.
PA5-20269 can be used with blocking peptide PEP-0384.
Bcl-2 related Protein A1 is a member of the BCL-2 protein family. The proteins of this family form hetero- or homodimers and act as anti- and pro-apoptotic regulators that are involved in a wide variety of cellular activities such as embryonic development, homeostasis and tumorigenesis. The protein encoded by this gene is able to reduce the release of pro-apoptotic cytochrome c from mitochondria and block caspase activation. This gene is a direct transcription target of NF-kappa B in response to inflammatory mediators, and has been shown to be up-regulated by different extracellular signals, such as granulocyte-macrophage colony-stimulating factor (GM-CSF), CD40, phorbol ester and inflammatory cytokine TNF and IL-1, which suggests a cytoprotective function that is essential for lymphocyte activation as well as cell survival.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A1-A; B-cell leukemia/lymphoma 2 related protein A1; B-cell leukemia/lymphoma 2 related protein A1a; B-cell leukemia/lymphoma 2 related protein A1d; Bcl-2-like protein 5; Bcl-2-related protein A1; Bcl2-L-5; BCL2-related protein A1; BCL2-related protein A1d; GRS protein; hematopoietic BCL2-related protein A1; Hemopoietic-specific early response protein; Protein BFL-1; Protein GRS
Gene Aliases: A1; ACC-1; ACC-2; ACC1; ACC2; BB218357; BCL2A1; Bcl2a1a; Bcl2a1d; BCL2L5; Bfl-1; BFL1; GRS; HBPA1
UniProt ID: (Human) Q16548, (Mouse) Q07440
Entrez Gene ID: (Human) 597, (Rat) 170929, (Mouse) 12044
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support