Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Description: This 5E7 monoclonal antibody recognizes mouse B lymphocyte-induced maturation protein-1 (Blimp-1), which is also known as Prdm1. Blimp-1 is a transcriptional repressor with five zinc finger domains. Blimp-1 is expressed by terminally differentiating B and T cells. Blimp-1 is also expressed by some dendritic cells, and some NK cells. This transcriptional repressor is required for B cell differentiation into plasma cells and serves as a critical antagonist of CD4+ follicular helper T cell differentiation. While Blimp-1 is not required for priming and development of T and B cell effector function, it is critical for effector cell terminal differentiation. Blimp-1 promotes effector molecule secretion while suppressing proliferation. Blimp-1 also acts as a transcriptional antagonist of Bcl-6 and inhibits genes that are related to memory cell differentiation.
Applications Reported: This 5E7 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.
Applications Tested: This 5E7 antibody has been tested by intracellular staining followed by flow cytometric analysis of stimulated mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00) and protocol. Please refer to "Staining Intracellular Antigens for Flow Cytometry, Protocol B: One step protocol for intracellular (nuclear) proteins" located at Flow Protocols. This may be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser
Blimp-1 (B lymphocyte maturation protein-1) is a nuclear protein with five zinc fingers in its carboxy terminus. It is a transcriptional repressor that is both required and sufficient to trigger terminal differentiation of B lymphocytes and monocyte/macrophages. Blimp-1 is expressed in, and may be required for terminally differentiated epithelial cells, retinal neurons and other lineage cells. It has also been implicated in the repression of beta-interferon (IFN-beta) and c-myc gene expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: B lymphocyte-induced maturation protein 1; Beta-interferon gene positive regulatory domain I-binding factor; BLIMP 1; Blimp-1; MGC118922; MGC118923; MGC118924; MGC118925; OTTHUMP00000016918; OTTHUMP00000202173; PR domain zinc finger protein 1; PR domain-containing protein 1; PRDI BF1; RP1-134E15.1
Gene Aliases: b2b1765Clo; Blimp-1; Blimp1; PRDI-BF1; Prdm1; ZNFPR1A1
UniProt ID: (Mouse) Q60636
Entrez Gene ID: (Mouse) 12142
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support