Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GRLVQTAAQQ VAEDKFVFDL PDYESINHVV VFMLGTIPFP EGMGGSVYFS YPDSNGMPVW QLLGFVTNGK PSAIFKI
Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Required for organization and/or function of the secretory apparatus in Clara cells in lung.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lethal gene on chromosome 7 Rinchik 6 protein; lethal, Chr 7, Rinchik 6; Protein Hikeshi
Gene Aliases: 0610007P06Rik; 1110002N09Rik; C11orf73; HIKESHI; HLD13; HSPC138; HSPC179; HSPC248; l(7)6Rn; L7RN6; OPI10
UniProt ID: (Human) Q53FT3, (Rat) Q5M808, (Mouse) Q9DD02
Entrez Gene ID: (Human) 51501, (Rat) 293103, (Mouse) 67669
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support