Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
C14orf1 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C14orf1; c14orf1-like protein; chromosome unknown open reading frame, human C14orf1; ergosterol biosynthetic protein 28; probable ergosterol biosynthetic protein 28; putative ergosterol biosynthetic protein 28; putative membrane protein; similar to yeast Yer044c; unnamed protein product
Gene Aliases: 0610007P14Rik; 1190004E09Rik; C14orf1; C20H14orf1; C24H14orf1; C8H14orf1; I79_009437; NET51; ORF11; RGD1310769
UniProt ID: (Human) Q9UKR5, (Mouse) Q9ERY9
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support