Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VVGRMQTMVT EQAAQGSSAP DLRVQYLRQI HANHETLLRE VQTLRDRLCT EDEASSCATA QRLLQVYRQL RHPSLILL
FAAP100 is a component of the Fanconi anemia (FA; MIM 277650) core complex and is required for core complex stability and FANCD2 (see MIM 227646) monoubiquitination (Ling et al., 2007 [PubMed 17396147]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fanconi anemia associated protein 100 kDa subunit; Fanconi anemia core complex 100 kDa subunit; Fanconi anemia core complex-associated protein 100; Fanconi anemia-associated protein of 100 kDa; Fanconi anemia-associated protein, 100kDa
Gene Aliases: C17orf70; FAAP100
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support