Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Elongin BC and Polycomb repressive complex 2-associated protein (EPOP) also know as C17ORF96 is a scaffold protein that serves as a bridging partner between the PRC2/EED-EZH2 complex and the elongin BC complex. It is required to fine tune the transcriptional status of polycomb group (PcG) target genes in embryonic stem cells (ESCs). C17ORF96 also plays a key role in genomic regions that display both active and repressive chromatin properties in pluripotent stem cells by sustaining low level expression of PcG target genes. It acts by recruiting the elongin BC complex, thereby restricting excessive activity of the PRC2/EED-EZH2 complex. C17ORF96 also has interacts with USP7 and promotes deubiquitination of H2B at promoter sites.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Elongin BC and Polycomb repressive complex 2-associated protein; Embryonic stem cell-specific PRC2 subunit p48; ES cell-specific PRC2 subunit p48; uncharacterized protein C17orf96 homolog
Gene Aliases: AA409164; AI413509; AW539173; E13; Epop; esPRC2p48
UniProt ID: (Mouse) Q7TNS8
Entrez Gene ID: (Mouse) 103551
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support