Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Antibody detects endogenous levels of total CS061.
This gene encodes a T cell adaptor protein, a class of intracellular molecules with modular domains capable of recruiting additional proteins but that exhibit no intrinsic enzymatic activity. The encoded protein contains a unique N-terminal region followed by a PH domain and C-terminal SH3 domain. Along with the adhesion and degranulation-promoting adaptor protein, the encoded protein plays a critical role in inside-out signaling by coupling T-cell antigen receptor stimulation to the activation of integrins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nonsense-mediated mRNA decay factor SMG9; protein smg-9 homolog; protein SMG9
Gene Aliases: 1500002O20Rik; AI429693; C19orf61; F17127_1; HBMS; N28092; RGD1309888; SMG9
UniProt ID: (Human) Q9H0W8, (Mouse) Q9DB90, (Rat) Q5PQS6
Entrez Gene ID: (Human) 56006, (Mouse) 71997, (Rat) 365215
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support