Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGKIALQLKA TLENITNLRP VGEDFRWYLK MKCGNCGEIS DKWQYIRLMD SVALKGGRGS ASMVQKCKLC ARENSIEILS STIKPYNAED NENFKTIVEF ECRGLEPVDF QPQAGFAAEG VESGTAFSDI NLQEKDWTDY DEKAQESVGI YEVTHQFVKC
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CXXC motif containing zinc binding protein; UPF0587 protein C1orf123
Gene Aliases: C1orf123; CZIB
UniProt ID: (Human) Q9NWV4
Entrez Gene ID: (Human) 54987
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support