Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKYLTVGDLE DCLLNKVDLR RQQISQAVEE VQKVVHHLTT NISNQDIRFQ AVPYSDTYNE NIKVLAPSQF LVTVPIKGLA GYREAREQHW RYYTLQGTRL PCPLRDPEGL QQWLEVEQFM KSLWQWHETD VNIDGDIVPA KVLLVFRKLV ENAVRTCHLS GKVSLLGNRS AVWVAVETSA YQVELELVPA VEIPTTWSKK ARWPRCLQRW PSQERVECIK SFGFNLLACS NYHWQLSFLR AEQVLLEQLD EDGGCRRKCF QVMRHLKEDI WCPGNRPVIT SHHLQTVLFW TCEKYPHFKD WQVFSKAFLR LVRKLHKCVS QHFLKHYFVR NSNLFQCTNP TELDTVAQKL ATFLKNPQIG PP
MAB21L3 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mab-21-like 3; Protein mab-21-like 3
Gene Aliases: C1orf161; MAB21L3
UniProt ID: (Human) Q8N8X9, (Mouse) Q8CI17
Entrez Gene ID: (Human) 126868, (Mouse) 242125
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support