Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji.
Predicted reactivity: Mouse (97%), Rat (95%), Rhesus Monkey (100%), Bovine (96%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C1orf165, also known as CFAP299 (Cilia and Flagella Associated Protein 299), is located on chromosome 1 and encodes a protein that is implicated in ciliary function and structure. CFAP299 is essential for the proper assembly and motility of cilia and flagella, structures that are vital for various cellular processes, including cell motility, fluid movement, and signal transduction. Mutations in CFAP299 have been linked to primary ciliary dyskinesia (PCD), which is characterized by chronic respiratory tract infections, abnormal positioning of internal organs (situs inversus), and infertility due to defective cilia and flagella movement. The protein has been shown to interact with other ciliary proteins and play a role in the regulation of dynein arms and microtubule structures necessary for ciliary beating.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BEN domain-containing protein 5
Gene Aliases: BEND5; C1orf165
UniProt ID: (Human) Q7L4P6
Entrez Gene ID: (Human) 79656
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support