Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lymphocyte expansion molecule; uncharacterized protein C1orf177; uncharacterized protein C1orf177 homolog
Gene Aliases: C1orf177; LEM; LEXM
UniProt ID: (Mouse) A2AVQ5, (Human) Q3ZCV2, (Rat) B1H283
Entrez Gene ID: (Mouse) 242602, (Human) 163747, (Rat) 500516
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support